Cytomegalovirus (CMV) CMV pp65 Peptide
Supplier: Anaspec Inc
AS-63937
103007-816EA
187.08
USD
103007-816
Cytomegalovirus (CMV) CMV pp65 Peptide
Proteins and Peptides
This peptide is derived from amino acid residues 13 to 27 of the 65k lower matrix phosphoprotein of the human cytomegalovirus.It contains a nine-amino-acid sequence (LGPISGHVL) that matches the consensus binding motif for a major histocompatibility complex H2-Dd T-cell epitope.
Sequence:SVLGPISGHVLKAVF
MW:1523.8 Da
% peak area by HPLC:95
Storage condition:-20° C
Sequence:SVLGPISGHVLKAVF
MW:1523.8 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...