CSK tide, FAM (Carboxyfluorescein)
Supplier: Anaspec Inc
AS-63842
103007-770EA
423.85
USD
103007-770
CSK tide, FAM (Carboxyfluorescein)
Proteins and Peptides
This is a FAM labeled peptide substrate (Abs/Em = 494/521 nm) for C-terminal Src kinase (Csk) and many other kinases such as Axl, cKit, ERBB4, Fes, Flt3, IGF-1 R, MET, MUSK, PYK2, Ret, TIE2, TrkA, VEGF-R1 and VEGF-R2.
Sequence:5-FAM-KKKKEEIYFFFG-NH2
MW:1921.2 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...