Myosin Regulatory Light Chain MRCL3 (11-24)
Supplier: Anaspec Inc
AS-63803
103007-738EA
173.92
USD
103007-738
Myosin Regulatory Light Chain MRCL3 (11-24)
Proteins and Peptides
This is a peptide substrate for myosin light chain kinase (MLCK). The sequence is derived from amino acid residues 11-24 of human myosin regulatory light chain MRCL3.
Sequence:KKRPQRATSNVFAM-NH2
MW:1633 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...