BDC2.5 Mimotope
Supplier: Anaspec Inc
AS-63774
103007-718EA
184.15
USD
103007-718
BDC2.5 Mimotope
Proteins and Peptides
The TCR transgenic model (BDC2.5) mimitope was used in type 1 diabetes (T1D) study. T1D is an autoimmune disease in which T cells mediate damage to pancreatic islet beta cells. T1D is caused by autoreactive T cell destruction of insulin-producing cells. BDC2.5 mimotope was utilized to support the study on antigen presentation of antigenic peptides to islet autoantigen-specific T cells.
Sequence: RTRPLWVRME
MW: 1343.6 Da
% Peak area by HPLC: 95
Storage condition: -20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...