Human Insulin beta Chain Peptide (15-23)
Supplier: Anaspec Inc
AS-63773
103007-716EA
146.15
USD
103007-716
Human Insulin beta Chain Peptide (15-23)
Proteins and Peptides
This is amino acids 15 to 23 fragment of the insulin beta chain recognized by islet-associated T cells. This peptide is also known as INS. It was used in diabetes studies to assay the IFN-beta and IL-17 production by diabetogenic CD4 or CD8 T cells. This peptide stains the INS-reactive CTL clone, but doesn’t stain the splenic CD8+ T cells from NOD or 8.3-TCR-transgenic NOD mice.
Sequence: LYLVCGERG
MW: 1009.2 Da
% Peak area by HPLC: 95
Storage condition: -20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...