3 x Hemagglutinin (HA) Tag
Supplier: Anaspec Inc
AS-63764
103007-714EA
651.85
USD
103007-714
3 x Hemagglutinin (HA) Tag
Proteins and Peptides
This tag peptide may be used to detect proteins and peptides, and to facilitate functional analysis of proteins of interest.
Sequence:MEYPYDVPDYAAEYPYDVPDYAAEYPYDVPDYAAKLE
MW:4372.7 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...