ClearPoint™ Human beta-Amyloid (1-42)
Supplier: Anaspec Inc
AS-63738
103007-700EA
286.46
USD
103007-700
ClearPoint™ Human beta-Amyloid (1-42)
Proteins and Peptides
ClearPoint™ beta-Amyloid (1-42) is a heavy-isotope labeled peptide. All the Arginine and Lysines have universally labeled 13C and 15N.
Sequence: DAEF-R*-HDSGYEVHHQ-K*-LVFFAEDVGSN-K*-GAIIGLMVGGVVIA [R*=R(U-13C6, U-15N4) & K*=K(U-13C6, U-15N2)]
Molecular Weight: 4540.1 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...