Human FRETS-VWF73 (Fluorescence-Quenching Substrate for ADAMTS-13)
Supplier: Anaspec Inc
AS-63728-01
AS-63728-05
103007-686EA
644.54
USD
103007-686
103007-688
Human FRETS-VWF73 (Fluorescence-Quenching Substrate for ADAMTS-13)
Proteins and Peptides
FRETS-VWF73 is a very powerful Fluorescence-Quenching Substrate for the VWF cleaving protease ADAMTS-13. It is commonly used as a useful tool for the rapid measurement of ADAMTS-13 activity in plasma and is a predictive marker for various thrombotic diseases like TPP. Our new substrate FRETS-VWF73 can be easily used with a 96-well format in commercial plate readers.
Sequence: DRE-Dap(Nma)-APNLVYMVTG-Dpa-PASDEIKRLPGDIQVVPIGVGPNANVQELERIGWPNAPILIQDFETLPREAPDLVLQR
MW: 8314.3 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...