[Lys(Ac)5]-Histone H4 (1-20)
Supplier: Anaspec Inc
AS-63683
103007-660EA
339.08
USD
103007-660
[Lys(Ac)5]-Histone H4 (1-20)
Proteins and Peptides
This is a histone 4 peptide acetylated at lysine 5. This form of histone acetylated lysine is found to be enriched at the 5' ends of the coding regions and is known to enhance gene expression.
Sequence:SGRG-K(Ac)-GGKGLGKGGAKRHRK
MW:2034.4 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...