[Lys(Me2)4]-Histone H3 (1-21)

Supplier: Anaspec Inc

AS-63677
103007-648EA 423.85 USD
103007-648
[Lys(Me2)4]-Histone H3 (1-21)
Proteins and Peptides

This is a dimethylated lysine at position 4 of the 1-21 amino acid histone H3 peptide. This form of histone methylation is characterized as being enriched around transcription start sites (TSS). It has been observed that a subgroup of genes linked to T cell functions showed high levels of H3K4me2 within their gene body. While enrichment for H3K4me2 within the gene body is a characteristic trait of expressed genes in yeast, in contrast, H3K4me2 has been shown to be predominantly enriched around the TSS in mammals.
Sequence:ART-K(Me2)-QTARKSTGGKAPRKQLA
MW:2282.6 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR