Human Beta-Amyloid (1-55)
Supplier: Anaspec Inc
AS-63335
103007-612EA
442.85
USD
103007-612
Human Beta-Amyloid (1-55)
Proteins and Peptides
Beta-amyloid 1-55 is the the 672-726 fragment of APP.
Sequence: DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIATVIVITLVMLKKK
Molecular Weight: 5982.1 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...