Human Beta-Amyloid (11-42), HiLyte™ Fluor 488
Supplier: Anaspec Inc
AS-63327
103007-610EA
339.08
USD
103007-610
Human Beta-Amyloid (11-42), HiLyte™ Fluor 488
Proteins and Peptides
This is amino acids 11 to 42 fragment of b-amyloid peptide labeled with HiLyte™ Fluor 488, Abs/Em=503/528 nm. Post-mortem Alzheimer’s diseased (AD) brain specimens reveal significant levels of this b -amyloid peptide within the insoluble amyloid pools. HiLyte™ Fluor 488-labeled b-amyloid (11-42) has a brighter intensity than FITC or FAM-labeled b-amyloid (11-42).
Sequence: HiLyte™ Fluor 488-EVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA
Molecular Weight: 3692.3 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...