Human Protease Activated Receptor 3
Supplier: Anaspec Inc
AS-62938
103007-560EA
134.46
USD
103007-560
Human Protease Activated Receptor 3
Proteins and Peptides
This peptide is one of the four PAR sub-types, and is acted upon by thrombin; not trypsin. It is a high-affinity thrombin receptor. PAR-3 mRNA is expressed in the cutaneous mast cells of humans. Co-expression of PAR3 and PAR4 enhances thrombin action suggesting that PAR3 alone does not mediate transmembrane signaling but instead functions as a cofactor to activate PAR4.
Sequence: SFNGGP-NH2
MW: 576.6 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...