Human;Mouse;Rat Beta-Amyloid (16-22)
Supplier: Anaspec Inc
AS-62897
103007-544EA
146.15
USD
103007-544
Human;Mouse;Rat Beta-Amyloid (16-22)
Proteins and Peptides
This is a short fragment of the b-Amyloid peptide containing two aromatic phenylalanine residues. Short peptide models have provided novel insight into the mechanistic issues of amyloid formation. This heptapeptide fragment has the capacity of forming typical amyloid fibrils in vitro. Aromatic interactions are important in many cases of amyloid formation.
Sequence: KLVFFAE
Molecular Weight: 853 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...