Shepherdin (79–87)
Supplier: Anaspec Inc
AS-62748
103007-504EA
146.15
USD
103007-504
Shepherdin (79–87)
Proteins and Peptides
This is amino acids 79 to 87 fragment of shepherdin, a novel peptidomimetic antagonist of the complex between Hsp90 and survivin, another key regulator of tumor cell viability. For its potent and broad antitumor activity, selectivity of action in tumor cells versus normal tissues, and inhibition of tumor growth in vivo without toxicity, shepherdin may offer a promising approach for rational cancer therapy. This sequence is also known as K79–K90.
Sequence:KHSSGCAFL
MW:949.1 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...