MBP (84-105)
Supplier: Anaspec Inc
AS-62739
103007-498EA
339.08
USD
103007-498
MBP (84-105)
Proteins and Peptides
This peptide is a fragment of myelin basic protein (MBP), which was used in multiple sclerosis studies. It corresponds to amino acids 85-106 of the guinea pig sequence, and amino acids 86-107 of the human protein.
Sequence:VVHFFKNIVTPRTPPPSQGKGR
MW:2462.9 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...