40Gap 27, Connexin Mimetic
Supplier: Anaspec Inc
AS-62641
103007-448EA
157.85
USD
103007-448
40Gap 27, Connexin Mimetic
Proteins and Peptides
This peptide corresponds to the GAP27 domain of the second extracellular loop of dominant vascular connexin (Cx40), designated as 40Gap 27. It was used to investigate mechanisms through which oxidant stress impairs communication via gap junctions. When administered, 40Gap27 attenuates endothelium-dependent subintimal smooth muscle hyperpolarization.
Sequence:SRPTEKNVFIV
MW:1289.5 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...