WRW4 Peptide
Supplier: Anaspec Inc
AS-62638
103007-444EA
146.15
USD
103007-444
WRW4 Peptide
Proteins and Peptides
This peptide inhibits binding of one of formyl peptide receptor-like 1 (FPRL1) agonists WKYMVm to its specific receptor. FPRL1 is an important classical chemoattractant receptor that is expressed in phagocytic cells in the peripheral blood and brain. Activation of FPRL1 is closely related to inflammatory responses in the host defense mechanism and neurodegenerative disorders.
Sequence:WRWWWW
MW:1105.3 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...