Caloxin 3A1
Supplier: Anaspec Inc
AS-62606
103007-420EA
279.15
USD
103007-420
Caloxin 3A1
Proteins and Peptides
This peptide belongs to caloxins, the extracellular plasma membrane (PM) Ca2+ pump inhibitors. Caloxin 3A1 inhibits plasma membrane calcium pumps (PMCAs) but not the sarcoplasmic reticulum Ca2+-pump. This peptide does not inhibit formation of the acylphosphate intermediate from ATP. Related peptides: Caloxin 1A1 (cat# 62605) and Caloxin 2A1 (cat# 62604).
Sequence:WSSTSSVSAPLEFGGGGSAK
MW:1912.1 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...