Caloxin 2A1
Supplier: Anaspec Inc
AS-62604
103007-418EA
216.31
USD
103007-418
Caloxin 2A1
Proteins and Peptides
Caloxins are extracellular plasma membrane (PM) Ca2+ pump inhibitors. Caloxin 2A1 is a PM Ca2+ pump inhibitor selectively binding to an extracellular domain. It inhibits Ca2+-Mg2+-ATPase in human erythrocyte leaky ghosts. Caloxin 2A1 is active at an extracellular site, the peptide can simply be added exogenously to inhibit the plasma membrane calcium ATPase (PMCA). Related peptides: Caloxin 1A1 (cat# 62605) and Caloxin 3A1 (cat# 62606).
Sequence:VSNSNWPSFPSSGGG
MW:1479.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...