Lymphocytic Choreomeningitis Virus (LCMV) 276-286
Supplier: Anaspec Inc
AS-62539
103007-390EA
182.69
USD
103007-390
Lymphocytic Choreomeningitis Virus (LCMV) 276-286
Proteins and Peptides
This peptide is amino acids 276 to 286 fragment of the lymphocytic choriomeningitis virus (LCMV) glycoprotein (GP), also known as GP276. It is the H-2Db restricted epitope. LCMV has been routinely exploited for the study of adaptive immune responses to viral infection. Fifty to seventy percent of CD8 T cells at the peak of LCMV infection appear to be specific for five LCMV-derived epitopes including GP276.
Sequence:SGVENPGGYCL
MW:1095.2 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...