Human;Mouse;Rat Biotin-LC-beta-Amyloid (22-41), Biotin
Supplier: Anaspec Inc
AS-62455
103007-352EA
371.23
USD
103007-352
Human;Mouse;Rat Biotin-LC-beta-Amyloid (22-41), Biotin
Proteins and Peptides
This is amino acids 22 to 41 fragment of beta-amyloid peptide, biotinylated through an LC spacer at the N-terminus.
Sequence: Biotin-LC-EDVGSNKGAIIGLMVGGVVI
Molecular Weight: 2267.8 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...