Tetanus Toxin (830–844)
Supplier: Anaspec Inc
AS-62410
103007-326EA
194.38
USD
103007-326
Tetanus Toxin (830–844)
Proteins and Peptides
This peptide belongs to 830 to 844 amino acid sequence of the tetanus toxin Tc, human, common for most MHC molecules.
Sequence:QYIKANSKFIGITEL
MW:1725 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...