Cyclo [-RGDyK(HiLyte™ Fluor 750)]
Supplier: Anaspec Inc
AS-62333-01
103007-284EA
571.46
USD
103007-284
Cyclo [-RGDyK(HiLyte™ Fluor 750)]
Proteins and Peptides
Cyclo(-RGDy-K) is labeled with HiLyte™ Fluor 750 (Abs/Em=754/778 nm). This cyclic RGD peptide contains a subsituted d-Tyr instead of d-Phe on the fourth position. Substitution with Tyr results in a high affinity and selectivity for the avb3 integrin. Substitution with Tyr also allows for electrophilic radiohalogenation if desired.
Sequence:Cyclo[-RGDy-K(Hilyte Fluor™ 750)]
MW:1630.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...