Influenza Matrix Protein (62-70)
Supplier: Anaspec Inc
AS-62288
103007-276EA
157.85
USD
103007-276
Influenza Matrix Protein (62-70)
Proteins and Peptides
This peptide is a fragment of the influenza virus matrix protein amino acid residues 62 to 70. It contains the core sequence of a new major histocompatibility complex class II-restricted T-cell epitope of influenza virus matrix protein. This epitope was detected in the peripheral blood of influenza A patients.
Sequence:GFVFTLTVPSER
MW:1352.6 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...