Noxa BH3, Peptide 1
Supplier: Anaspec Inc
AS-62282
103007-274EA
371.23
USD
103007-274
Noxa BH3, Peptide 1
Proteins and Peptides
This peptide belongs to the Bcl-2 family of proteins. Noxa gene encodes a Bcl-2 homology 3 (BH3)-only member of this family; it contains the BH3 region, not other BH domains. When ectopically expressed, Noxa undergoes BH3 motif-dependent localization to mitochondria, it interacts with anti-apoptotic Bcl-2 family members resulting in the activation of caspase-9.
Sequence:PAELEVECATQLRRFGDKLNFRQKLL
MW:3075.6 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...