HIV TAT-NSF700 Fusion Peptide
Supplier: Anaspec Inc
AS-62238
103007-258EA
612.38
USD
103007-258
HIV TAT-NSF700 Fusion Peptide
Proteins and Peptides
This peptide is the N-Ethyl-maleimide-sensitive factor (NSF) inhibitor fusion polypeptide composed of 11-amino acid cell permeable HIV transactivating regulatory protein (TAT) domain fused to a 22 amino acid NSF domain. TAT-NSF700 inhibits thrombin-induced exocytosis of endothelial cells in a dose-responsive manner.
Sequence: YGRKKRRQRRR-GGG-LLDYVPIGPRFSNLVLQALLVL
MW: 4167 Da
% Peak area by HPLC: 95
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...