Human Beta-Amyloid (1-40)
Supplier: Anaspec Inc
AS-62233
103007-256EA
230.92
USD
103007-256
Human Beta-Amyloid (1-40)
Proteins and Peptides
This synthetic peptide corresponds to ß--Amyloid (1-40) with an additional cysteine at the C-terminus.
Sequence: DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVC
Molecular Weight: 4433 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...