Human [Gly21]-beta-Amyloid (1-40)
Supplier: Anaspec Inc
This peptide is the mutant form of the b-Amyloid peptide (1-40). The mutation within the coding region of the ß-Amyloid precursor protein (APP) results in substitution of glycine for alanine in this peptide. Presenile dementia is present in a pattern consistent in the family of British origin with the dominant inheritance of Flemish APP mutation. The impact of the point mutation A21G on b-Amyloid structure and dynamics varies from b-Amyloid (1-40) to b-Amyloid (1-42).
Sequence: DAEFRHDSGYEVHHQKLVFFGEDVGSNKGAIIGLMVGGVV
Molecular Weight: 4315.9 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...