Gamma-Fibrinogen (377-395)
Supplier: Anaspec Inc
AS-62129
103007-202EA
225.08
USD
103007-202
Gamma-Fibrinogen (377-395)
Proteins and Peptides
This is a scrambled sequence of the g-fibrinogen amino acids 377 to 395. The original fibrinogen-derived inhibitory peptide attenuates microglia activation and suppresses relapsing paralysis in multiple sclerosis (MS). This scrambled peptide fails to produce these functions.
Sequence:KMMISYTFPIERTGLISNK
MW:2229.7 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...