Apidaecin IB
Supplier: Anaspec Inc
AS-62044
103007-152EA
216.31
USD
103007-152
Apidaecin IB
Proteins and Peptides
Apidaecin IB is an insect antimicrobial peptide showing a significant sequence homology and a common mechanism of action with drosocin, but is devoid of any pore-forming activity. Apidaecins are the most prominent components of the honey bee humoral defense against microbial invasion.
Sequence:GNNRPVYIPQPRPPHPRL
MW:2108.4 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...