390 MMP FRET Substrate V, Mca-DNP
Supplier: Anaspec Inc
Matrix Metalloproteinases (MMPs) are a large family of endopeptidases. Collectively, MMPs can degrade all kinds of extracellular matrix proteins, and can also process a number of bioactive molecules. They are known to be involved in the cleavage of cell surface receptors, the release of apoptotic ligands, and chemokine/cytokine inactivation. MMPs are also thought to play a major role in cell behaviors such as cell proliferation, migration (adhesion/dispersion), differentiation, angiogenesis, apoptosis, and host defense.
This peptide is a substrate for the MMP-12, macrophage elastase, Abs/Em = 325/393 nm. MMPs are considered to be critical mediators of both normal and pathological tissue remodeling processes.
Sequence:Mca-PLGLEEA-Dap(Dnp)-NH2
MW:1193.2 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...