Human;Mouse;Rat Beta-Amyloid (25-35)
Supplier: Anaspec Inc
AS-61971
103007-120EA
146.15
USD
103007-120
Human;Mouse;Rat Beta-Amyloid (25-35)
Proteins and Peptides
This is the scrambled beta-Amyloid peptide amino acids 25 to 35. Pairing this peptide with the native b-Amyloid 25 to 35 amino acids peptide has been used to recognize its structure and functions.
Sequence: MAKGINGISGL
Molecular Weight: 1060.3 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...