Renin 520 Peptide
Supplier: Anaspec Inc
AS-61872-01
103007-076EA
415.08
USD
103007-076
Renin 520 Peptide
Proteins and Peptides
This FRET peptide is a specific substrate for renin. This renin peptide substrate may be used for screening of renin inhibitors. In the FRET peptide, the fluorescence of 5-FAM is quenched by QXL® 520. Upon cleavage into two separate fragments by renin, the fluorescence of 5-FAM is recovered, and can be monitored at excitation/emission = 490/520 nm. This substrate is employed in the SensoLyte® 520 Renin Assay Kit, cat # AS-72040 from AnaSpec.
MW: 2000 - 2200 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...