Cathepsin S Substrate, AMC (7-amino-4-methylcoumarin)
Supplier: Anaspec Inc
AS-61859
103007-070EA
239.69
USD
103007-070
Cathepsin S Substrate, AMC (7-amino-4-methylcoumarin)
Proteins and Peptides
Cathepsins are a class of globular lysosomal proteases, playing a vital role in mammalian cellular turnover. They degrade polypeptides and are distinguished by their substrate specificities. Cathepsin S is a cysteine proteinase involved in the pathogenesis of autoimmune diseases, atherosclerosis, cancer, obesity and related diseases.
This peptide is a cathepsin S substrate fluorescently labeled with AMC (Ex/Em=354 nm/442 nm). It can be used to measure cathepsin S activity.
Sequence:Ac-KQKLR-AMC
MW:871.1 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...