Histone Deacetylase Substrate, AMC (7-Amino-4-Methylcoumarin)
Supplier: Anaspec Inc
AS-61855
103007-068EA
279.15
USD
103007-068
Histone Deacetylase Substrate, AMC (7-Amino-4-Methylcoumarin)
Proteins and Peptides
This novel fluorogenic substrate of HDACs was synthesized with an epsilon-acetylated lysyl moiety and an adjacent MCA moiety at the C-terminus of the peptide chain. The assay utilizing this substrate provides a good tool to characterize the HDAC activity. HDACs are important enzymes for the transcriptional regulation of gene expression.
Sequence:Ac-RGK(Ac)-AMC
MW:600.7 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...