Human Beta-Amyloid (13-28)
Supplier: Anaspec Inc
AS-61802
103006-994EA
194.38
USD
103006-994
Human Beta-Amyloid (13-28)
Proteins and Peptides
This β-Amyloid peptide 13 to 27 amino acid residues was used to study the kinetics of β-amyloid formation.
Sequence: HHQKLVFFAEDVGSNK
Molecular Weight: 1856.1 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...