Histone H1-derived Peptide, FAM (Carboxyfluorescein)
Supplier: Anaspec Inc
AS-61762
103006-970EA
434.08
USD
103006-970
Histone H1-derived Peptide, FAM (Carboxyfluorescein)
Proteins and Peptides
This is a FAM-labeled Histone H1-derived peptide (Ab/Em = 494/521 nm). Histone H1-derived peptide phosphorylated by protein kinase A, is a substrate for CDK2 and CDK5 (cyclin dependent kinase 5) and PKA.
Sequence:5-FAM-GGGPATPKKAKKL
MW:1610.8 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...