Histone H1-derived Peptide
Supplier: Anaspec Inc
AS-61761
103006-968EA
160.77
USD
103006-968
Histone H1-derived Peptide
Proteins and Peptides
Histone H1-derived peptide, phosphorylated by protein kinase A, is a substrate for CDK2 and CDK5 (cyclin dependent kinase 5) and PKA.
Sequence:GGGPATPKKAKKL
MW:1252.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...