Abltide
Supplier: Anaspec Inc
AS-61749
103006-960EA
194.38
USD
103006-960
Abltide
Proteins and Peptides
Abltide is a peptide substrate for Abl Kinase (Abl protein tyrosine kinase), a partner in the gag-Abl fusion protein of the Abelson murine leukemia virus. Used in Western blot and kinase assay.
Sequence:KKGEAIYAAPFA-NH2
MW:1264.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...