RS domain derived peptide
Supplier: Anaspec Inc
AS-61722
103006-942EA
146.15
USD
103006-942
RS domain derived peptide
Proteins and Peptides
This peptide is a substrate for Clk/Sty and is phosphorylated by Clk/Sty protein kinase (Km=102 microM).
Sequence:GRSRSRSRSR
MW:1204.3 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...