Bid BH3 Peptide
Supplier: Anaspec Inc
AS-61711
103006-922EA
239.69
USD
103006-922
Bid BH3 Peptide
Proteins and Peptides
BID is a pro-apoptotic member of the 'BH3-only' (BOPS) subset of the BCL-2 family of proteins that constitute a critical control point in apoptosis. Bid is the first of the BOPs reported to bind and activate Bcl-2, Bax, and Bak. Bid serves as a death-inducing ligand that moves from the cytosol to the mitochondrial membrane to inactivate Bcl-2 or to activate Bax.
Sequence:EDIIRNIARHLAQVGDSMDR
MW:2309.6 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...