Histone H3 (116–136)
Supplier: Anaspec Inc
AS-61704
103006-916EA
323
USD
103006-916
Histone H3 (116–136)
Proteins and Peptides
This peptide spans the C-terminus of histone H3, amino acids 116 to 136.
Sequence:KRVTIMPKDIQLARRIRGERA
MW:2508 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...