Histone H3 (1-21), Biotin
Supplier: Anaspec Inc
AS-61702
103006-912EA
432.62
USD
103006-912
Histone H3 (1-21), Biotin
Proteins and Peptides
This sequence corresponds to the first 21 amino acids of the NH2 terminal of histone H3 followed by a GG linker and a biotinylated lysine. This peptide was used to investigate the characteristics and mechanisms of ethanol-induced histone H3 acetylation in rat hepatocytes. Immunocytochemical and immunoblot analyses revealed that ethanol treatment significantly increased H3 acetylation at Lys9 with negligible effects at Lys14, 18, and 23.
Sequence:ARTKQTARKSTGGKAPRKQLA-GGK(Biotin)-NH2
MW:2723.2 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...