Human Protease Activated Receptor 1
Supplier: Anaspec Inc
AS-61530
103006-810EA
130.08
USD
103006-810
Human Protease Activated Receptor 1
Proteins and Peptides
This protease-activated receptor-1 (PAR-1) selective peptide induces cyclooxygenase-2 (COX-2) protein and mRNA expression in human endothelial cells. Studies show that intratracheal instillation of the PAR1-specific peptide increases lung edema during high-tidal-volume ventilation.
Sequence: TFLLRN
MW: 762.9 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...