PDKtide
Supplier: Anaspec Inc
AS-64225
103006-682EA
701.54
USD
103006-682
PDKtide
Proteins and Peptides
This peptide is a substrate for PDK1 (Phosphatidylinositide-Dependent Kinase 1).
Sequence:KTFCGTPEYLAPEVRREPRILSEEEQEMFRDFDYIADWC
MW:4771.4 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...