Human Myelin oligodendrocyte glycoprotein
Supplier: Anaspec Inc
AS-64218
103006-678EA
371.23
USD
103006-678
Human Myelin oligodendrocyte glycoprotein
Proteins and Peptides
This peptide sequence is found in residues 89 to 113 of human MOG (Myelin Oligodendrocyte Glycoprotein). Studies suggest this is an HLA-DR2 restricted MOG epitope.
Sequence: RFSDEGGFTCFFRDHSYQEEAAMEL
MW: 2973.2 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...