Rat Atrial Natriuretic Peptide (4-18)

Supplier: Anaspec Inc

AS-62839
103006-666EA 279.15 USD
103006-666
Rat Atrial Natriuretic Peptide (4-18)
Proteins and Peptides

This peptide corresponds to amino acids 4 to 18 of rat atrial natriuretic peptide (ANP). It has been used as a cystein containing peptide model in quantitative mass spectroscopic analysis. This fragment is also related to C-ANP (4-23);  (Des-Gln18,des-Ser19,des-Gly20,22,des-Leu21), which is completely selective in discriminating rat C-AMP receptors.
Sequence:RSSCFGGRIDRIGAC-NH2 (Disulfide bridge 4-15)
MW:1594.9 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR