p53 (17-26)
Supplier: Anaspec Inc
AS-61375
103006-594EA
146.15
USD
103006-594
p53 (17-26)
Proteins and Peptides
This peptide is amino acids 17 to 26 fragment of p53, the Mdm-2 binding domain of p53 known also as p53N. This sequence contains all of the residues that contact the binding domain of Mdm-2. The tumor suppressor protein p53 is important in maintaining genome stability and in preventing cancer development.
Sequence:ETFSDLWKLL
MW:1251.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...