Human MUC5AC
Supplier: Anaspec Inc
AS-61329
103006-578EA
194.38
USD
103006-578
Human MUC5AC
Proteins and Peptides
This peptide is derived from the human mucin MUC5AC gene sequence. Data suggest that MUC5A and MUC5C are part of the same gene MUC5AC, which is distinct from MUC5B. The gene MUC5AC is mainly expressed in gastric, tracheo-bronchial mucosae and some tumors, it exhibits two kinds of deduced peptide domains, one of which is 8 amino acid tandemly repeated domain, a consensus peptide TTSTTSAP.
Sequence:GTTPSPVPTTSTTSAP
MW:1501.6 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...